Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253119.2 | XP_006253181.1 | 5 - 6 | 770 | 59 | |
XM_006253120.2 | XP_006253182.1 | 5 - 6 | 648 | 59 | ||
GenBank mRNA | BC081703.1 | - | 5 - 6 | 727 | 59 | |
FQ215471.1 | - | 6 - 7 | 894 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
