Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053650.1 | NP_446102.1 | 3 - 4 | 404 | 77 | |
XM_006253120.2 | XP_006253182.1 | 3 - 4 | 440 | 77 | ||
GenBank mRNA | AF002281.1 | - | 3 - 4 | 422 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
