Assay Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253167.2 | XP_006253229.1 | 8 - 9 | 1130 | 142 | |
XM_341433.6 | XP_341434.3 | 8 - 9 | 1127 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
