Assay Details
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771266.1 | XP_008769488.1 | 6 - 7 | 1257 | 116 | |
XM_008771267.1 | XP_008769489.1 | 6 - 7 | 1177 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
