Assay Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013882.2 | NP_001013904.2 | 1 - 2 | 204 | 90 | |
NM_001161512.1 | NP_001154984.1 | 1 - 2 | 420 | 90 | ||
GenBank mRNA | BC087138.1 | - | 1 - 2 | 159 | 7125992 | 90 |
BP504069.1 | - | 422 | 90 | |||
DY317307.1 | - | 1 - 2 | 172 | 90 | ||
EV776012.1 | - | 1 - 2 | 229 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
