Assay Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013882.2 | NP_001013904.2 | 4 - 5 | 554 | 102 | |
GenBank mRNA | BC087138.1 | - | 4 - 5 | 509 | 7125992 | 102 |
DY317307.1 | - | 4 - 5 | 522 | 102 | ||
EV776012.1 | - | 4 - 5 | 579 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
