Assay Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161512.1 | NP_001154984.1 | 291 | 72 | ||
GenBank mRNA | BP504069.1 | - | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001161512.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
