Assay Details
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770708.1 | XP_008768930.1 | 10 - 11 | 1883 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
