Assay Details
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108866.1 | NP_001102336.1 | 4 - 5 | 716 | 131 | |
XM_006251995.2 | XP_006252057.1 | 4 - 5 | 921 | 131 | ||
XM_006251996.2 | XP_006252058.1 | 4 - 5 | 712 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
