Assay Details
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108866.1 | NP_001102336.1 | 5 - 6 | 822 | 80 | |
XM_006251995.2 | XP_006252057.1 | 5 - 6 | 1027 | 80 | ||
XM_006251996.2 | XP_006252058.1 | 5 - 6 | 818 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
