Assay Details
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108866.1 | NP_001102336.1 | 6 - 7 | 1005 | 61 | |
XM_006251995.2 | XP_006252057.1 | 6 - 7 | 1210 | 61 | ||
XM_006251996.2 | XP_006252058.1 | 6 - 7 | 1001 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
