Assay Details
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013036.1 | NP_001013054.1 | 1 - 2 | 67 | 119 | |
XM_006251925.2 | XP_006251987.1 | 1 - 2 | 128 | 119 | ||
XM_006251926.2 | XP_006251988.1 | 1 - 2 | 128 | 119 | ||
XM_006251927.2 | XP_006251989.1 | 1 - 2 | 149 | 119 | ||
GenBank mRNA | BC089848.1 | - | 1 - 2 | 67 | 119 | |
FQ211507.1 | - | 1 - 2 | 89 | 119 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC089848.1, NM_001013036.1, XM_006251925.2, XM_006251926.2, XM_006251927.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
