Assay Details
Gene Symbol: Khnyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751468.3 | XP_003751516.1 | 7 - 8 | 2026 | 66 | |
XM_006252032.2 | XP_006252094.1 | 7 - 8 | 2221 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
