Assay Details
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017240.2 | NP_058936.1 | 9 - 10 | 1011 | 87 | |
XM_006251951.2 | XP_006252013.1 | 11 - 12 | 1161 | 87 | ||
GenBank mRNA | X15939.1 | - | 9 - 10 | 1013 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
