Assay Details
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053442.1 | NP_445894.1 | 4 - 5 | 1236 | 106 | |
GenBank mRNA | AB024400.1 | - | 4 - 5 | 1236 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
