Assay Details
Gene Symbol: Mrpl52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108375.2 | NP_001101845.2 | 2 - 3 | 108 | 89 | |
XM_006251976.2 | XP_006252038.1 | 2 - 3 | 133 | 89 | ||
GenBank mRNA | BC166922.1 | - | 2 - 3 | 108 | 6919316 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
