Assay Details
Gene Symbol: Mrpl52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108375.2 | NP_001101845.2 | 1 - 2 | 47 | 82 | |
GenBank mRNA | BC166922.1 | - | 1 - 2 | 47 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166922.1, NM_001108375.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
