Assay Details
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001078405.6 | XP_001078405.4 | 8 - 9 | 812 | 91 | |
XM_008770715.1 | XP_008768937.1 | 8 - 9 | 828 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
