Assay Details
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013995.1 | NP_001014017.1 | 1 - 2 | 154 | 69 | |
GenBank mRNA | BC079109.1 | - | 1 - 2 | 154 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079109.1, NM_001013995.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
