Assay Details
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013995.1 | NP_001014017.1 | 5 - 6 | 635 | 70 | |
XM_006251955.1 | XP_006252017.1 | 5 - 6 | 607 | 70 | ||
XM_006251956.2 | XP_006252018.1 | 5 - 6 | 786 | 70 | ||
GenBank mRNA | BC079109.1 | - | 5 - 6 | 635 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
