Assay Details
Gene Symbol: Haus4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013995.1 | NP_001014017.1 | 9 - 10 | 1075 | 112 | |
XM_006251955.1 | XP_006252017.1 | 9 - 10 | 1047 | 112 | ||
XM_006251956.2 | XP_006252018.1 | 9 - 10 | 1226 | 112 | ||
GenBank mRNA | BC079109.1 | - | 9 - 10 | 1075 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
