Assay Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031341.1 | NP_112631.1 | 8 - 9 | 1448 | 81 | |
XM_006252020.2 | XP_006252082.1 | 8 - 9 | 1384 | 81 | ||
XM_006252021.2 | XP_006252083.1 | 11 - 12 | 1853 | 81 | ||
XM_006252023.2 | XP_006252085.1 | 8 - 9 | 1389 | 81 | ||
XM_006252024.2 | XP_006252086.1 | 8 - 9 | 2078 | 81 | ||
XM_008770705.1 | XP_008768927.1 | 10 - 11 | 1796 | 81 | ||
GenBank mRNA | AB020520.1 | - | 8 - 9 | 1448 | 81 | |
AF200684.1 | - | 7 - 8 | 1256 | 81 | ||
BC091142.1 | - | 8 - 9 | 1363 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
