Assay Details
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC078685.1 | - | 1 - 2 | 81 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078685.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
