Assay Details
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175757.2 | NP_786933.1 | 5 - 6 | 689 | 96 | |
XM_006252058.2 | XP_006252120.1 | 3 - 4 | 1210 | 96 | ||
GenBank mRNA | AY040223.1 | - | 5 - 6 | 689 | 96 | |
BC078685.1 | - | 5 - 6 | 663 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
