Assay Details
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106039.1 | NP_001099509.1 | 3 - 4 | 324 | 73 | |
XM_008770675.1 | XP_008768897.1 | 3 - 4 | 1452 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
