Assay Details
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007621.1 | NP_001007622.1 | 8 - 9 | 1178 | 65 | |
XM_006251946.2 | XP_006252008.1 | 8 - 9 | 1043 | 65 | ||
XM_006251947.2 | XP_006252009.1 | 9 - 10 | 1111 | 65 | ||
XM_006251948.2 | XP_006252010.1 | 9 - 10 | 1089 | 65 | ||
GenBank mRNA | BC079263.1 | - | 8 - 9 | 1178 | 65 | |
FQ211258.1 | - | 6 - 7 | 889 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
