Assay Details
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007621.1 | NP_001007622.1 | 3 - 4 | 662 | 70 | |
XM_006251946.2 | XP_006252008.1 | 3 - 4 | 527 | 70 | ||
XM_006251947.2 | XP_006252009.1 | 4 - 5 | 595 | 70 | ||
XM_006251948.2 | XP_006252010.1 | 4 - 5 | 573 | 70 | ||
GenBank mRNA | BC079263.1 | - | 3 - 4 | 662 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
