Assay Details
Gene Symbol: Ngdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107263.1 | NP_001100733.1 | 1 - 2 | 61 | 58 | |
XM_008770676.1 | XP_008768898.1 | 1 - 2 | 1046 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107263.1, XM_008770676.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
