Assay Details
Gene Symbol: Ngdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107263.1 | NP_001100733.1 | 4 - 5 | 333 | 142 | |
XM_006251966.2 | XP_006252028.1 | 4 - 5 | 526 | 142 | ||
XM_008770676.1 | XP_008768898.1 | 4 - 5 | 1318 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
