Assay Details
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017239.2 | NP_058935.2 | 15 - 16 | 1999 | 86 | |
GenBank mRNA | X15938.1 | - | 15 - 16 | 1996 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
