Assay Details
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109505.1 | NP_001102975.1 | 1 - 2 | 27 | 61 | |
XM_006231758.2 | XP_006231820.1 | 1 - 2 | 43 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109505.1, XM_006231758.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
