Assay Details
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109505.1 | NP_001102975.1 | 6 - 7 | 578 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21414 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P17958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21414 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21414 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P17958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21414 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95274 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10785 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51224 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85413 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27556 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85732 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75667 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59179 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24305 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27417 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64044 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76579 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44644 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20339 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10820317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820321 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10820316 | - | - |
Rat Genome 230 2.0 Array | 1376971_at | - | - |
Rat Genome 230 2.0 Array | 1376971_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376971_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376971_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200017976.rn.1 | TC0200002583.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200017977.rn.1 | TC0200002583.rn.1 | RTA TC |
Rat Expression Set 230 | 1376971_at | - | - |
Rat Expression Set 230 | 1376971_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10820316 | - | - |
Rat Genome U34 Set | rc_AI072626_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21566406 | - | - |
