Assay Details
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ103710.1 | - | 10 - 11 | 1128 | 72 | |
L20319.1 | - | 10 - 11 | 1249 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P638181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40588 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P638181 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P638181 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P638181 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P638181 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37045 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69405 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85051 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54929 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89100 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72641 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10812480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812472 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812473 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812471 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812479 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812478 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812481 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812475 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10812470 | - | - |
Rat Genome 230 2.0 Array | 1386979_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386979_at | - | - |
Affymetrix HT Rat Focus Array | 1386979_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386979_at | - | - |
Rat Expression Set 230 | 1386979_at | - | - |
Rat Expression Set 230 | 1386979_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812470 | - | - |
Rat Genome U34 Set | rc_AI012275_at | - | - |
