Assay Details
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ103710.1 | - | 6 - 7 | 797 | 96 | |
L20319.1 | - | 6 - 7 | 918 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
