Assay Details
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ103710.1 | - | 8 - 9 | 1022 | 132 | |
L20319.1 | - | 8 - 9 | 1143 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
