Assay Details
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030850.1 | NP_110477.1 | 1 - 2 | 98 | 67 | |
GenBank mRNA | AF038870.1 | - | 1 - 2 | 98 | 67 | |
BC078811.1 | - | 1 - 2 | 85 | 7130736 | 67 | |
U96133.1 | - | 1 - 2 | 87 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF038870.1,BC078811.1,NM_030850.1,U96133.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
