Assay Details
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031593.1 | NP_113781.1 | 2 - 3 | 799 | 63 | |
GenBank mRNA | AF060174.1 | - | 2 - 3 | 799 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
