Assay Details
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 6 - 7 | 861 | 57 | |
XM_008760635.1 | XP_008758857.1 | 6 - 7 | 1429 | 57 | ||
GenBank mRNA | AB007688.1 | - | 6 - 7 | 868 | 57 | |
AB017140.1 | - | 6 - 7 | 861 | 57 | ||
AF093268.1 | - | 6 - 7 | 1298 | 57 | ||
AJ276328.1 | - | 6 - 7 | 581 | 57 | ||
AY189943.1 | - | 2 - 3 | 240 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
