Assay Details
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 1 - 2 | 296 | 110 | |
XM_006231775.2 | XP_006231837.1 | 1 - 2 | 877 | 108 | ||
GenBank mRNA | AB003726.1 | - | 1 - 2 | 672 | 110 | |
AB007688.1 | - | 1 - 2 | 303 | 110 | ||
AB017140.1 | - | 1 - 2 | 296 | 110 | ||
AF030088.2 | - | 1 - 2 | 1207 | 110 | ||
AF093267.1 | - | 1 - 2 | 741 | 110 | ||
AF093268.1 | - | 1 - 2 | 733 | 110 | ||
U92079.1 | - | 1 - 2 | 568 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB003726.1,AB007688.1,AB017140.1,AF093267.1,AF093268.1,NM_031707.1,U92079.1,XM_006231775.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
