Assay Details
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231775.2 | XP_006231837.1 | 5 - 6 | 1403 | 66 | |
GenBank mRNA | AF093267.1 | - | 5 - 6 | 1267 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
