Assay Details
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199192.1 | NP_001186121.1 | 3 - 4 | 256 | 142 | |
GenBank mRNA | DN934981.1 | - | 3 - 4 | 282 | 142 | |
FM121157.1 | - | 3 - 4 | 228 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() |
