Assay Details
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199192.1 | NP_001186121.1 | 4 - 5 | 318 | 96 | |
GenBank mRNA | DN934981.1 | - | 4 - 5 | 344 | 96 | |
FM121157.1 | - | 4 - 5 | 290 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
