Assay Details
Gene Symbol: Ppwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106406.1 | NP_001099876.1 | 5 - 6 | 1096 | 112 | |
XM_006231887.2 | XP_006231949.1 | 5 - 6 | 873 | 112 | ||
XM_008760666.1 | XP_008758888.1 | 7 - 8 | 868 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
