Assay Details
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077428.1 | NP_001070896.1 | 12 - 13 | 887 | 112 | |
XM_006231844.2 | XP_006231906.1 | 12 - 13 | 969 | 112 | ||
XM_006231845.2 | XP_006231907.1 | 12 - 13 | 1127 | 112 | ||
GenBank mRNA | BC126097.1 | - | 12 - 13 | 887 | 112 | |
FQ229200.1 | - | 828 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
