Assay Details
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077428.1 | NP_001070896.1 | 6 - 7 | 377 | 135 | |
XM_006231844.2 | XP_006231906.1 | 6 - 7 | 459 | 135 | ||
XM_006231845.2 | XP_006231907.1 | 6 - 7 | 617 | 135 | ||
GenBank mRNA | BC126097.1 | - | 6 - 7 | 377 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
