Assay Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033443.1 | NP_254278.1 | 1 - 2 | 392 | 148 | |
GenBank mRNA | BC168241.1 | - | 363 | 148 | ||
CK482953.1 | - | 1 - 2 | 392 | 148 | ||
D49434.1 | - | 1 - 2 | 134 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
