Assay Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033443.1 | NP_254278.1 | 4 - 5 | 985 | 76 | |
GenBank mRNA | BC127450.1 | - | 4 - 5 | 876 | 76 | |
BC168241.1 | - | 956 | 76 | |||
CF113828.1 | - | 1 - 2 | 153 | 76 | ||
D49434.1 | - | 4 - 5 | 727 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
