Assay Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033443.1 | NP_254278.1 | 5 - 6 | 1215 | 82 | |
GenBank mRNA | BC127450.1 | - | 5 - 6 | 1106 | 82 | |
BC168241.1 | - | 1186 | 82 | |||
CF113828.1 | - | 2 - 3 | 383 | 82 | ||
D49434.1 | - | 5 - 6 | 957 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
