Assay Details
Gene Symbol: Hcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053375.1 | NP_445827.1 | 5 - 6 | 1414 | 129 | |
GenBank mRNA | AF247450.1 | - | 5 - 6 | 1414 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
