Assay Details
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105953.2 | NP_001099423.2 | 2 - 3 | 144 | 95 | |
NM_001277263.1 | NP_001264192.1 | 2 - 3 | 221 | 95 | ||
GenBank mRNA | BC169086.1 | - | 1 - 2 | 111 | 95 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC169086.1, NM_001105953.2, NM_001277263.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
